Bulb pipettes, one mark, Class A
Artikelnr. HIRS134G15
Lieferant: HIRSCHMANN
Spezifikationen
- Kapazität:15 ml
- Bezeichnung:Unserialised pipettes
- Seq:03
- VE:6
- Toleranz:0,03 ± ml
Spezifikationen
Über diesen Artikel
Borosilicate glass.
- Designed to deliver according to ASTM Specification E969, Class A requirements
- Colour-coded for ease in sorting and selection
- Fine black enamel capacity line and markings
- Serialised pipettes include a Certificate of Identification and Accuracy
Certifications: ASTM E969