ContactUsLinkComponent
Sampling probe assemblies for autosamplers, AS 93 Plus/S10
Sampling probe assemblies for autosamplers, AS 93 Plus/S10
Catalog # PERCB3001770
Supplier:  PerkinElmer
Sampling probe assemblies for autosamplers, AS 93 Plus/S10
Catalog # PERCB3001770
Supplier:  PerkinElmer
Specifications
  • Für:
    AS-90/90A/90 Plus/91/93 Plus autosamplers
  • Bezeichnung:
    Sampling probe, 0,6 mm Ø int.
  • Seq:
    01
  • VE:
    1