Cleaning cloths, Praktitex® Pro
Artikelnr. ZVGA10294-01
Lieferant: Zellstoff-Vertriebs-GmbH & Co. KG
Spezifikationen
- Verpackt:10 wipes/pack
- Farbe:Pink
- L×B:400×400 mm
- VE:10
Spezifikationen
Über diesen Artikel
This microfibre cloth (80% polyester, 20% polyamide) has an open structure and is therefore ideal for cleaning heavily fouled surfaces.
- Extremely fine fibres penetrate deep into the surface and thus effectively loosen adhering and grease-containing fouling
- Washable and therefore reusable
Weight: 300 g/m²