Nitrites test kits, NI-15
Artikelnr. HACH2182000
Lieferant: Hach
CAS-Nummer:
Spezifikationen
- VE:1 KIT
- Bezeichnung:Nitrite test kit, LR, NI-15
- Messbereich:0 - 0,40 mg/l NO₂- N
Spezifikationen
Über diesen Artikel
The accurate visual test kit method to determine nitrogen as nitrite by diazotisation of sample followed by colour comparison.
- Simple and cost effective
- Portable test kit
Delivery: Kit includes colour disc, comparator box and tubes, reagents ( but not deionised water), instructions and carrying case.