Hydrogen sulfide test kit, HS-C
Catalog # HACH2537800
Supplier: Hach
CAS Number:
Specifications
- Chemische Methode:Effervescence of H2S
- VE:1 KIT
- Bezeichnung:Hydrogen sulphide test kit, HS-C
- Messbereich:0 to 5 mg/l H₂S
Specifications
About this item
The kit is used to determine sulphide pollution of sewage water, waste water and other sources resulting from decomposition of organic matter. In the HS-C kit, sulphide discolours a detection paper. The results can be obtained by comparison of the colour to a printed chart.
- Reliable, cost effective colour comparison method
Delivery information: Includes reaction vial, effervescent tablets, test papers, comparator chart and instructions, in a cardboard box.