AuftragsverfolgungKontakt
Insulation stripper | Avantor
Insulation stripper
Insulation stripper, chrome plated, handles insulated with multi-component grips
Artikelnr. 238-5410
Anbieter:   KNIPEX WERKE
Insulation stripper
Artikelnr. 238-5410
Anbieter:  KNIPEX WERKE
Insulation stripper
Insulation stripper
Insulation stripper, chrome plated, handles insulated with multi-component grips
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Chrome plated, forged, oil-hardened insulation stripper.
  • For single, multiple and fine stranded conductors with plastic or rubber insulation max. Ø 5,0 mm or respectively 10 mm² cable cross section
  • Easy adjustment to the required diameter of solid or stranded wire with knurled screw and lock nut
  • Handles insulated with multi-component grips
Zertifizierung: DIN EN 60900, IEC 60900. VDE-tested
Spezifikationen
  • Bezeichnung:
    Insulation stripper, chrome plated, handles insulated with multi-component grips