ContactUsLinkComponent
Swivel pad holder
Artikelnr. 129-0120
Lieferant:  PERFEX
undefined
Swivel pad holder
Artikelnr. 129-0120
Lieferant:  PERFEX
Spezifikationen
  • Farbe:
    Blau
  • Bezeichnung:
    Pad holder, Perfex 5000
  • Seq:
    02
  • VE:
    1
Spezifikationen
Über diesen Artikel
Swivel pad holder is designed to twist and turn for cleaning hard-to-reach areas. Its velcro base holds pads securely in place. Attach a polymer adjustable handle to the pad holder to clean floors, baseboards, walls, windows, and ceilings. Also, ideal for cleaning fine surfaces, including sinks, vinyl, and stainless steel.
Ordering information: pads, and handle are sold separately, please contact us for details.