Floor maintainer, Carefree Stride 1000
Catalog # DIVY454500
Supplier: Diversey
Specifications
- Bezeichnung:Carefree Stride 1000, floor maintainer, neutral detergent
- Verpackt:2 × 5 l
- VE:2 Can
Specifications
About this item
Carefree Stride is a neutral, low foam detergent which is suitable for all polished (protected) and unprotected floors. It can be used in either scrubber drier or manual cleaning methods.
- Excellent emulsification of soil, fragrance-free
- Fast, effective removal of everyday dirt and black heel marks