Mops, SuperMop AG and SuperMop Micro AG
Artikelnr. VILE100273
Lieferant: FHP VILEDA SCS
Spezifikationen
- Bezeichnung:Steel SuperMop handle, coated and painted, with monoclick connector for SuperMop fitting, 100273
- Länge:1380 mm
- Seq:11
- VE:1
Spezifikationen
Über diesen Artikel
SuperMop Ag and SuperMop Micro Ag are the top hygienic solutions for fast, effective and intensive cleaning of small and medium sized floor areas. The unique construction that hinders the growth and multiplication of bacteria, now has silver ions added that inhibits 99,99% of bacteria growth within the mop.
- Independently tested to inhibit bacteria growth
- Oval design, allows easy cleaning of corners
- High resistance to detergents and bleaches
- Low friction, lightweight non-woven-material
- Highly absorbent for spill pick up
- Colour-coded in four colours to avoid cross contamination