Sumps for small containers, PE-EP
Sump for storing small containers, with 100 l sump capacity, PE-100-EP25, Rot, 680×750×420 mm
Artikelnr. LACOP70-7083-A
Anbieter: LACONT UMWELTTECHNIK
Springe zu:
Product Details & Documents
Ideal for the storage of water-endangering and aggressive media.
- Rotationally-moulded body of red PE
- Circumferential stabilising ribs
- Equipped with protective cover
Spezifikationen
- Farbe:Rot
- Bezeichnung:Sump for storing small containers, with 100 l sump capacity, PE-100-EP25
- B×T×H:680×750×420 mm