Reinraum-Wischtücher, getränkt, AlphaSat® mit Vectra® AlphaSorb® 10
Catalog # TEXWTX1057
Supplier: Texwipe
Specifications
- Verpackt:40 Tücher/wiederverschließbaren Beutel, 12 Beutel/Karton
- Bezeichnung:TX1057
- L×B:230×230 mm
- VE:480
Specifications
About this item
AlphaSat® mit Vectra® AlphaSorb® 10 sind ultrasaubere AlphaSorb 10 Tücher (100% Polyester) mit Kantenversiegelung und getränkt mit 10% Isopropylalkohol in Halbleiterqualität (0,2 µm gefiltert). Verpackt in einer benutzerfreundlichen Flat-Pack-Verpackung.
- Superior strength and abrasion resistance in a continuous-filament knitted substrate
- Low particle generation and low extractable levels help prevent product and cleanroom contamination
- Eliminates problems associated with purchasing, storing, filtering and diluting alcohol
- Reduces alcohol consumption and eliminates over-spray of alcohol
Ideal for cleaning production equipment and environmental surfaces for reduction of particulate contamination, for wipedown of articles to be passed or carried into cleanrooms, for process chamber wipedowns in semiconductor processing and for cleaning fingerprints and light soils from sensitive medical devices prior to packaging and terminal sterilisation.
Packaging: Packaged in a cleanroom environment. Wipes are protected in a sealed outer bag for contamination-free introduction into the cleanroom.
Caution: *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904.