Attachable screwthread tubes
GL 18
Artikelnr. SMIT746/GL18
Anbieter: SMITH SCIENTIFIC
Springe zu:
Product Details & Documents
Borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
DIN screw thread GL.
Spezifikationen
- Gewinde:GL 18
- Seq:02