Mini hip joint with cross-section
Catalog # BINH1000168
Supplier: 3B Scientific
Specifications
- Bezeichnung:Mini hip joint with cross-section
- VE:1
Specifications
About this item
The mini hip joint model has been reduced to a half of natural size but has kept all of the functionality of larger hip joint models. In addition to the external anatomical structures, the medical or teaching professional now has the possibility to explain what is happening from 'within' the hip joint.
- Mounted on the base
Dimensions: 16×12×20 cm
Weight: 0,2 kg