Stand for spinal columns and skeletons, 3 part
Artikelnr. BINH1000132
Lieferant: 3B Scientific
Spezifikationen
- Bezeichnung:Stand for spinal columns and skeletons, 3 part
- VE:1
Spezifikationen
Über diesen Artikel
New, worldwide unique spine and skeleton stand. This stand works great for spines and hanging skeleton models.
- Can be placed on the floor
- Or mounted on a wall
- Made of nickel-plated steel
Stand measures 86,36 cm tall on a 24,13 cm square base
Weight: 0,75 kg