Micro spoon and spatula sampling set, SP Bel-Art
Artikelnr. BELAF367050000
Lieferant: Bel-Art Products, a Part of SP
Spezifikationen
- Bezeichnung:Sampling set, includes two micro spoons, two V-shaped spatula, eight tools with flat spatulas ends (6 square and 2 rounded ends) and one pair of blunt tip tweezers in a carrying case
- VE:1 KIT
Spezifikationen
Über diesen Artikel
This kit includes six different, small size, double-ended tools and a pair of tweezers. Each double-ended tool has two different end shapes providing 12 implements to better match user needs. Kits include:
- Two small spoons for transferring and collecting both dry and liquid material
- Two V-shaped spatula ends of different sizes that can be used for lifting or spill-free carrying of material
- Eight flat spatulas ends (six square ends and two rounded) of various widths and lengths ideal for mixing, scraping and spreading
- Blunt tip tweezers have serrated inside surfaces for secure sample handling and grooved handles for better gripping
All tools are polished stainless steel for long life and easy cleaning and can be steam autoclaved at 121 °C. A carrying case protects and organises all tools when not in use.