Barrier tips, GEN3™, Axygen®
Artikelnr. 732-7515
Lieferant: Corning
Spezifikationen
- Verpackt:50 racks, 100 each
- Steril:+
- Volumen:100 - 1000 µl
- Seq:04
- Spitzentyp:Standard
- VE:5.000
Spezifikationen
Über diesen Artikel
These multi-barrier filter pipette tips offer the ultimate in protection against cross contamination using tri-filter technology. These tips will prevent liquid or aerosol from passing through and will lock on contact assuring the valuable samples and research have not been compromised.
- Effectively separate liquid from the self-sealing barrier using an additional protective layer of conventional polyethylene filter material
- Proprietary filter design prevents possible PCR inhibition
- Maxymum Recovery® technology for low sample retention and high yield
- Hinged rack for convenience and ease of use
- Certified DNase-/RNase-free and non pyrogenic
- Certified human gDNA and PCR inhibition free