Pasteur pipettes, disposable
Pasteur pipettes, disposable, borosilicate glass, with cotton plug, tip length 127 mm, 7 mm, 229 mm, 250/pack, 4 packs/case
Catalog # BELC1277-10009
Marketsource
Supplier: BELLCO GLASS
Jump to:
Product Details & Documents
Made of borosilicate glass.
Suitable for the transfer of all infectious materials, culture media and for routine sampling.
Specifications
- Ext.Ø:7 mm
- Verpackt:250/pack, 4 packs/case
- Bezeichnung:Pasteur pipettes, disposable, borosilicate glass, with cotton plug, tip length 127 mm
- Länge:229 mm