Berufsschuhe, Clogs, Professional Chiro Clogs
The anti-static chiro clogs range of professional footwear made from PU.
- Textile lining and ventilation holes for a good foot climate
- Toe bar keeps the foot from slipping
- Shock absorber heel
The clogs are ideal for use in hospitals, hygiene areas, pharmaceutical industries and research laboratories. Ideal for high-sensitive areas.
Colours: White, green
: EN ISO 20347:2004
Frequently Bought Together