Pipette tips for Liquidator™ 96, BioClean™ LTS™
Lieferant: RAININ
Zertifikate
Über diesen Artikel
Racked tips for Liquidator™ 96 bench-top pipettor with LTS™.
- In 96-well format - sturdy and reusable
- Fast loading, good protection & autoclavable
- BioClean: 100% free of DNA, DNAse, RNAse, ATP, Pyrogens, PCR inhibitors, additives and bioactive contaminants
- Advanced LTS design: No deformations, no cracks, no flash