Kaliumchlorid 3 mol/l (3 N) in wässriger Lösung
Catalog # METT51350080
Supplier: Mettler - Toledo
CAS Number:
Specifications
- VE:6
- Seq:0006
Specifications
About this item
:
:74.55 g/mol
Density:1.13 g/cm³ (20 °C)
Storage Temperature:Raumtemperatur
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360