About this item
HiTrap Capto SP ImpRes chromatography column is packed with a strong cation exchange modern resin for high resolution, small-scale protein purification.
- Suitable for screening of selectivity, binding and elution conditions, and for small-scale purifications
- Sulphonate (SP) strong cation exchanger
- Optimized for intermediate and polishing protein purification
- Convenient HiTrap format for easy connection to a syringe, peristaltic pump, or chromatography system such as an ÄKTA system
HiTrap Capto SP ImpRes column is prepacked with Capto SP ImpRes resin. The HiTrap format is designed to offer quality and flexibility in process-scale applications. Scaling up allows for high resolution combined with quick flow rates.
This resin couples a sulphonate strong cation exchanger to a chemically modified, high-flow agarose matrix. The matrix provides greater particle rigidity without compromising pore size, which allows for high bed heights and purification of viscous samples at high flow rates.
The resin’s high binding capacity, high flow rate, and low back pressure process cycle times and improve productivity at process scale.
Specifications
- Catalog No:
- 17-5468-51
- 17-5468-55
- 29-4004-60
- Max. Druck:
- 5 bar
- 5 bar
- 5 bar
- Lagerung:
- 4...30 °C, 20% Ethanol, 0,2 M Sodium Acetate
- 4...30 °C, 20% Ethanol, 0,2 M Sodium Acetate
- 4...30 °C, 20% Ethanol, 0,2 M Sodium Acetate
Specifications
Related Information
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements