Anion exchange HPLC column, NUCLEOSIL® Anion II
Supplier: MACHEREY-NAGEL
Certificates
About this item
This column for analysis of inorganic anions has a strongly basic anion exchanger with an exchange capacity of 50 µeq/g.
- Based on robust spherical silica gel
- Recommended buffer concentration for separation of inorganic anions: 2 mmol/L phthalate
- Preferred method of detection: conductivity or negative UV detection
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).
Specifications
- Catalog No:
- MANA720094.40
- MANA721169.30
- pH-Bereich:
- 2 - 7,5
- 2 - 7,5