Repetitive pipette, manual, AutoRep™
Catalog # RAINAR-M2
Supplier: RAININ
Specifications
- Volumen:10 - 5000 µl
- Bezeichnung:AutoRep™ M, manueller Dispenser
- Modell:AutoRep™ M
- Spitze:Encode Spitzen; 0,5 - 50 ml
- VE:1
Specifications
About this item
This manual repeater pipette can be used with Rainin Encode™ or Brand® PD-Tip® syringes or compatible syringes from other manufacturers.
- Precise and accurate
- Saves time and effort, speeding up long pipetting serie
- Lightweight, ergonomic design virtually eliminates all forces that contribute to repetitive strain injuries experienced in multi-dispense pipetting