Additives
Appearance, Mix Ratio,Product | Appearance | Mix Ratio | Data Sheet | Comment | |||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
MED-4102-1 | White | 1 PART | Download MED-4102-1 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4102-2 | Black | 1 PART | Download MED-4102-2 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4102-3 | Red | 1 PART | Download MED-4102-3 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4102-4 | Orange | 1 PART | Download MED-4102-4 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4102-5 | Yellow | 1 PART | Download MED-4102-5 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4102-6 | Green | 1 PART | Download MED-4102-6 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4102-7 | Dark Blue | 1 PART | Download MED-4102-7 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4502-1 | White | 1 PART | Download MED-4502-1 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4502-2 | Black | 1 PART | Download MED-4502-2 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4502-3 | Red | 1 PART | Download MED-4502-3 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4502-4 | Orange | 1 PART | Download MED-4502-4 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4502-5 | Yellow | 1 PART | Download MED-4502-5 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4502-6 | Green | 1 PART | Download MED-4502-6 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4502-7 | Blue | 1 PART | Download MED-4502-7 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4800-1 | White | 1 PART | Download MED-4800-1 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4800-2 | Black | 1 PART | Download MED-4800-2 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4800-3 | Red | 1 PART | Download MED-4800-3 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4800-4 | Orange | 1 PART | Download MED-4800-4 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4800-5 | Yellow | 1 PART | Download MED-4800-5 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4800-6 | Green | 1 PART | Download MED-4800-6 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4800-7 | Dark Blue | 1 PART | Download MED-4800-7 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4900-1 | White | 1 PART | Download MED-4900-1 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4900-2 | Black | 1 PART | Download MED-4900-2 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4900-3 | Red | 1 PART | Download MED-4900-3 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4900-4 | Orange | 1 PART | Download MED-4900-4 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4900-5 | Yellow | 1 PART | Download MED-4900-5 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4900-6 | Green | 1 PART | Download MED-4900-6 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4900-7 | Dark Blue | 1 PART | Download MED-4900-7 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED1-4102-2 | Black | 1 PART | Download MED1-4102-2 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED1-4102-7 | Blue | 1 PART | Download MED1-4102-7 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED1-4502-2 | Black | 1 PART | Download MED1-4502-2 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED1-4502-7 | Blue | 1 PART | Download MED1-4502-7 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED1-4800-2 | Black | 1 PART | Download MED1-4800-2 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||
MED1-4800-7 | Blue | 1 PART | Download MED1-4800-7 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||
MED1-4900-2 | Black | 1 PART | Download MED1-4900-2 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED1-4900-7 | Blue | 1 PART | Download MED1-4900-7 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED2-4102 | White | 1 PART | Download MED2-4102 data sheet | 75% BaSO4, Barium Sulfate HCR Masterbatch for Radio Opacity | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED2-4102-6 | Green | 1 PART | Download MED2-4102-6 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED2-4502 | White | 1 PART | Download MED2-4502 data sheet | HCR Masterbatch, 75% BaSO4 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED2-4502-6 | Green | 1 PART | Download MED2-4502-6 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED2-4800 | White | 1 PART | Download MED2-4800 data sheet | LSR Masterbatch, 50% BaSO4 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED2-4800-6 | Green | 1 PART | Download MED2-4800-6 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||
MED2-4900 | White | 1 PART | Download MED2-4900 data sheet | 50% BaSO4, Barium Sulfate LSR Masterbatch For Radio Opacity | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED2-4900-6 | Light Green | 1 PART | Download MED2-4900-6 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED3-4102 | White | 1 PART | Download MED3-4102 data sheet | 75% TiO2, Titanium Dioxide Hcrmasterbatch | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED3-4102-8 | Violet | 1 PART | Download MED3-4102-8 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED3-4502 | White | 1 PART | Download MED3-4502 data sheet | HCR Masterbatch, 75% TiO2 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED3-4502-8 | Violet | 1 PART | Download MED3-4502-8 data sheet | Masterbatch to add color to HCR platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED3-4800 | White | 1 PART | Download MED3-4800 data sheet | LSR Masterbatch, 50% TiO2 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED3-4800-8 | Violet | 1 PART | Download MED3-4800-8 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||
MED3-4900 | White | 1 PART | Download MED3-4900 data sheet | 50% TiO2, Titanium Dioxide LSR Masterbatch | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED3-4900-8 | Violet | 1 PART | Download MED3-4900-8 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED4-4900 | White | 1 PART | Download MED4-4900 data sheet | Ammonium Bicarbonate LSR Foam Masterbatch | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED50-4800-1 | Translucent White | 1 PART | Download MED50-4800-1 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||
MED50-4800-2 | Translucent Black | 1 PART | Download MED50-4800-2 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||
MED50-4800-3 | Translucent Red | 1 PART | Download MED50-4800-3 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||
MED50-4800-4 | Translucent Orange | 1 PART | Download MED50-4800-4 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||
MED50-4800-5 | Translucent Yellow | 1 PART | Download MED50-4800-5 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||
MED50-4800-7 | Translucent Dark Blue | 1 PART | Download MED50-4800-7 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||
MED50-4900-1 | Translucent White | 1 PART | Download MED50-4900-1 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED50-4900-2 | Translucent Black | 1 PART | Download MED50-4900-2 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED50-4900-3 | Translucent Red | 1 PART | Download MED50-4900-3 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED50-4900-4 | Translucent Orange | 1 PART | Download MED50-4900-4 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED50-4900-5 | Translucent Yellow | 1 PART | Download MED50-4900-5 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED50-4900-6 | Translucent Green | 1 PART | Download MED50-4900-6 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED50-4900-7 | Translucent Dark Blue | 1 PART | Download MED50-4900-7 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED51-4800-2 | Black | 1 PART | Download MED51-4800-2 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||
MED51-4800-7 | Translucent Blue | 1 PART | Download MED51-4800-7 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||
MED51-4900-7 | Translucent Blue | 1 PART | Download MED51-4900-7 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED52-4900-6 | Translucent Green | 1 PART | Download MED52-4900-6 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED53-4900-8 | Translucent Violet | 1 PART | Download MED53-4900-8 data sheet | Masterbatch to add color to platinum cure systems | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
Adhesives/Sealants
Cure System, Rheology, Work Time, Durometer,Product | Cure System | Rheology | Work Time | Durometer | Data Sheet | Comment | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CF2-3521-2 | Platinum | Thixotropic / non-slump | 1 hours | 35 Type A | Download CF2-3521-2 data sheet | Fuel Resistance | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CF5-3521-2 | Platinum | Thixotropic / non-slump | 3.5 hours | 30 Type A | Download CF5-3521-2 data sheet | 50% M fluoro. Black and will RTV in 48 hrs | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-1142 | Oxime | Thixotropic / non-slump | N/A | 45 Type A | Download CV-1142 data sheet | For applications requiring a broader operating temperature range. Available in black and white. 200 psi (1.4 MPa), Unprimed lap shear. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-1143 | Oxime | Thixotropic / non-slump | N/A | 45 Type A | Download CV-1143 data sheet | Non-Slump | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2187 | Platinum | Flowable / Self Leveling | 3 hours | 35 Type A | Download CV-2187 data sheet | Tough, self leveling and good adhesion to various metals. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2287 | Platinum | Flowable / Self Leveling | 3.5 hours | 30 Type A | Download CV-2287 data sheet | Low / High Temperature, Flowable, Fast Cure | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2289 | Platinum | Flowable / Self Leveling | 30 minutes | 30 Type A | Download CV-2289 data sheet | Tough, self leveling and good adhesion to various metals. Recommended for broad temperature applications. 400 psi (2.8 MPa), primed (CF1-135) lap shear. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2289-1 | Platinum | Flowable / Self Leveling | 30 minutes | 34 Type A | Download CV-2289-1 data sheet | Pourable Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2289-2 | Platinum | Flowable / Self Leveling | 30 minutes | 30 Type A | Download CV-2289-2 data sheet | Pourable Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2500 | Platinum | Flowable / Self Leveling | 2 hours | 50 Type A | Download CV-2500 data sheet | Pourable, Optically Clear | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2500-2 | Platinum | Flowable / Self Leveling | 3 hours | 50 Type A | Download CV-2500-2 data sheet | Low Viscosity, Fast Cure | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2510 | Alkoxy | Flowable / Self Leveling | 4 hours | 45 Type A | Download CV-2510 data sheet | Low / High Temperature, Flowable | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2566 | Alkoxy | Flowable / Self Leveling | 3 hours | 55 Type A | Download CV-2566 data sheet | Pourable RTV Cure, * 1 )Primed Lap Shear 500 psi (3.4 MPa) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2567 | Alkoxy | Flowable / Self Leveling | 6.5 hours | 21 Type A | Download CV-2567 data sheet | Diluent for Tin Condensation CV Silicones | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2568 | Alkoxy | Flowable / Self Leveling | 4 hours | 50 Type A | Download CV-2568 data sheet | Long Worktime, Low Density* 1, )Primed Lap Shear 100 psi (0.69 MPa) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2680-12 | Platinum | N/A | N/A | N/A | Download CV-2680-12 data sheet | Low Outgassing version of R-2682-12. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2686-12 | Platinum | N/A | N/A | N/A | Download CV-2686-12 data sheet | Use for RFI and EMI shielding in electrical and space applications. For applications requiring shorter work times, easy clean-up, and consistent bond thickness. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2688-12 | Platinum | Thixotropic / Non-slump | N/A | N/A | Download CV-2688-12 data sheet | For applications requiring shorter work times, easy clean-up, and consistent bond thickness | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV1-1142 | Oxime | Flowable / Self Leveling | N/A | 30 Type A | Download CV1-1142 data sheet | Low viscosity and self leveling. Recommended for board temperature applications. 65 psi (0.45 Mpa) primed (SP-120), lap shear. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV1-1142-4 | Oxime | Flowable / Self Leveling | N/A | 35 Type A | Download CV1-1142-4 data sheet | Self-leveling, Built-in UV Tracer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV1-2289-1 | Platinum | Flowable / Self Leveling | 4 hours | 30 Type A | Download CV1-2289-1 data sheet | White, versatile adhesive with 3 hr pot life. Recommended for broad temperature applications. 450 psi (3.1 MPa), primed (CF1-135) lap shear. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV1-2566 | Alkoxy | Flowable / Self Leveling | 3 hours | 50 Type A | Download CV1-2566 data sheet | *1) Primed Lap Shear 650 psi (4.5 MPa) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV10-2500 | Platinum | Flowable / Self Leveling | 3 hours | 50 Type A | Download CV10-2500 data sheet | High Durometer, Optically Clear | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV10-2568 | Platinum | Flowable / Self Leveling | 3 hours | 40 Type A | Download CV10-2568 data sheet | Fast Cure, Low Density*,4 )Primed Lap Shear 175 psi (1.2 MPa) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV14-2500 | Platinum | Flowable / Self Leveling | N/A | 30 Type A | Download CV14-2500 data sheet | Primerless Adhesion | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV15-2500 | Platinum | Flowable / Self Leveling | 3 hours | 50 Type A | Download CV15-2500 data sheet | *4 )Primed Lap Shear 225 psi (1.6 MPa), Optically Clear | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV16-2500 | Platinum | Flowable / Self Leveling | 2 hours | 40 Type A | Download CV16-2500 data sheet | Low / High Temperature,*4 ) Primed Lap Shear 200 psi (4.1 MPa), Optically Clear | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV2-1142 | Oxime | Thixotropic / non-slump | N/A | 50 Type A | Download CV2-1142 data sheet | Non-slump and excellent for applications where minimal flow is desired. For applications requiring a broader operating temperature range. Available in black and white. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV2-2289-1 | Platinum | Flowable / Self Leveling | N/A | 30 Type A | Download CV2-2289-1 data sheet | White, versatile adhesive with short pot life, tack free in 20 hrs Recommended for broad temperature applications. 300 psi (2.1 MPa) primed (CF1-135) lap shear. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV2-2566 | Alkoxy | Thixotropic / non-slump | 2 hours | 55 Type A | Download CV2-2566 data sheet | *2) Primed Lap Shear 625 psi (4.3 MPa) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV2-2646 | Alkoxy | Flowable / Self Leveling | 2 hours | 75 Type A | Download CV2-2646 data sheet | 0.003 ohmcm at 200 C | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV3-1142 | Oxime | Thixotropic / non-slump | N/A | 45 Type A | Download CV3-1142 data sheet | Spot Bonding, Available in Black & White | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV3-2500 | Platinum | Flowable / Self Leveling | 3 hours | 40 Type A | Download CV3-2500 data sheet | Low Viscosity, Potting & Encapsulant, Optically Clear | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV4-2289-1 | Platinum | Thixotropic / non-slump | 30 minutes | 30 Type A | Download CV4-2289-1 data sheet | Non-flowable | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV7-1142-1 | Oxime | Flowable / Self Leveling | N/A | 40 Type A | Download CV7-1142-1 data sheet | Recommended for extended operation at elevated temperatures. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV7-2289-1 | Platinum | Flowable / Self Leveling | N/A | 30 Type A | Download CV7-2289-1 data sheet | Primerless Adhesion | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
EPM-2411-2 | Platinum | Thixotropic / non-slump | N/A | 20 Type A | Download EPM-2411-2 data sheet | Low volatility, Thixotropic, dispensable and 0.5 mm maximum slump. Shipped and stored frozen. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
EPM-2412 | Platinum | Flowable / Self Leveling | 20 minutes | 30 Type A | Download EPM-2412 data sheet | Mobile Devices, Displace | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
EPM-2461 | Platinum | Thixotropic / non-slump | 1 minutes | 30 Type A | Download EPM-2461 data sheet | Carbon filled. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
EPM-2462 | Platinum | Thixotropic / non-slump | 2.75 hours | 85 Type A | Download EPM-2462 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
FS-3730 | Acetoxy | Thixotropic / non-slump | N/A | 33 Type A | Download FS-3730 data sheet | 100%M Fluoro general all purpose adhesive for environments exposed to hydrocarbon fuels or solvents. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
FS-3730-11 | Acetoxy | Thixotropic / non-slump | N/A | 40 Type A | Download FS-3730-11 data sheet | Lap Shear after 7 days *1) 275 psi (1.9 MPa) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
FS-3730-2 | Acetoxy | Thixotropic / non-slump | N/A | 40 Type A | Download FS-3730-2 data sheet | Lap Shear after 7 days *3) 300 psi (2.1 MPa) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
FS-3775 | Acetoxy | Flowable / Self Leveling | N/A | 30 Type A | Download FS-3775 data sheet | High Temperature, Fuel Resistant | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
FS1-3730 | Acetoxy | Thixotropic / non-slump | N/A | 35 Type A | Download FS1-3730 data sheet | Bonds Aggressively | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
FS3-3730 | Acetoxy | Thixotropic / non-slump | N/A | 35 Type A | Download FS3-3730 data sheet | 100%M Fluoro general all purpose adhesive for environments exposed to hydrocarbon fuels or solvents. Translucent. 200 psi (1.4 MPa) lap shear. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
FS9-3521 | Platinum | Flowable / Self Leveling | 3 hours | 29 Type A | Download FS9-3521 data sheet | 100% F, For applications requiring fuel and solvent resistance | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-1000 | Acetoxy | Flowable / Self Leveling | N/A | 25 Type A | Download MED-1000 data sheet | Medium flow, high extrusion rate | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-1011 | Acetoxy | Flowable / Self Leveling | N/A | 25 Type A | Download MED-1011 data sheet | Low flow | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-1031 | Oxime | Thixotropic / non-slump | N/A | 35 Type A | Download MED-1031 data sheet | Non-acidic leaving group | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-1037 | Acetoxy | Thixotropic / non-slump | N/A | 30 Type A | Download MED-1037 data sheet | Low-slump, solvent-free translucent silicone | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-1040 | Acetoxy | Flowable / Self Leveling | N/A | 23 Type A | Download MED-1040 data sheet | High flow | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-1131 | Oxime | Thixotropic / non-slump | N/A | 35 Type A | Download MED-1131 data sheet | Non-acidic leaving group | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-1137 | Acetoxy | Thixotropic / non-slump | N/A | 30 Type A | Download MED-1137 data sheet | Low-slump, solvent-free translucent silicone | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-1511 | Acetoxy | Flowable / Self Leveling | N/A | 25 Type A | Download MED-1511 data sheet | Low flow | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-1540 | Acetoxy | Flowable / Self Leveling | N/A | 23 Type A | Download MED-1540 data sheet | High flow | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-2000 | Acetoxy | Flowable / Self Leveling | N/A | 25 Type A | Download MED-2000 data sheet | Medium flow, high extrusion rate | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED1-4013 | Platinum | Thixotropic / non-slump | N/A | 20 Type A | Download MED1-4013 data sheet | Quick cure, high extrusion rate | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED1-4213 | Platinum | Thixotropic / non-slump | 15 minutes | 20 Type A | Download MED1-4213 data sheet | Quick cure, high extrusion rate | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED2-4013 | Platinum | Thixotropic / non-slump | N/A | 15 Type A | Download MED2-4013 data sheet | Quick cure, high extrusion rate | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED2-4213 | Platinum | Thixotropic / non-slump | 15 hours | 15 Type A | Download MED2-4213 data sheet | Quick cure, high extrusion rate | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED3-4013 | Platinum | Thixotropic / non-slump | N/A | 20 Type A | Download MED3-4013 data sheet | Quick cure, high extrusion rate | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED3-4213 | Platinum | Thixotropic / non-slump | 60 minutes | 20 Type A | Download MED3-4213 data sheet | Quick cure, high extrusion rate | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-1130 | Oxime | Thixotropic / non-slump | N/A | 35 Type A | Download R-1130 data sheet | Non-slump. Adhesive for a variety of substrates on or near electronic components. 485 psi (3.6 MPa), primed (SP-120) lap shear. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-1140 | Acetoxy | Thixotropic / non-slump | N/A | 30 Type A | Download R-1140 data sheet | Non-slump. General all purpose adhesive for a variety of substrates. 625 psi (4.3 MPa), primed (SP-120) lap shear. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-1400 | Platinum | Thixotropic / non-slump | N/A | 19 Type A | Download R-1400 data sheet | Thixotropic, dispensable, and 0.5mm maximum slump. Shipped and stored frozen. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2141 | N/A | Flowable / Self Leveling | 1.5 hours | 40 Type A | Download R-2141 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2145 | Platinum | Flowable / Self Leveling | 15 minutes | 45 Type A | Download R-2145 data sheet | Primerless Adhesive;Gray, tough, fast cure adhesive. Will RTV. 145 psi (1.0 MPa), unprimed and 600 psi (4.1 MPa) primed (CF1-135) lap shear. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2160 | Platinum | Flowable / Self Leveling | 1.5 hours | 20 Type A | Download R-2160 data sheet | Iron Red, flowable legacy high performance elastomer at elevated temperatures. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2186 | Platinum | Flowable / Self Leveling | 2.5 hours | 30 Type A | Download R-2186 data sheet | Legacy general all purpose adhesive. 475 psi (3.3 MPa), primed (CF1-135) lap shear. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2550 | Alkoxy | Flowable / Self Leveling | 6 hours | 35 Type A | Download R-2550 data sheet | Resists breakdown at high temperatures. Translucent and 1 hr. pot life. Not recommended for deep section cures. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2560 | Alkoxy | Flowable / Self Leveling | 1 minutes | 55 Type A | Download R-2560 data sheet | Resists breakdown at high temperatures. Red and 1 hr. pot life. Not recommended for deep section cures. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2630 | Platinum | Flowable / Self Leveling | 15 hours | 60 Type A | Download R-2630 data sheet | Carbon filled. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2634 | Alkoxy | Thixotropic / non-slump | 3 hours | 80 Type A | Download R-2634 data sheet | 200 psi lap shear primed w/SP-121. Conductive at elevated temperatures. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2682-12 | Platinum | N/A | N/A | N/A | Download R-2682-12 data sheet | For applications requiring shorter processing times, easy clean-up, and consistent bond thickness. Adheres to silicone, and some metals or plastics. Sheet is 0.013 inches (0.3 mm) and cures on surfaces where the Activator is applied. Cures at room temperature or rapidly with heat. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R1-2145 | Platinum | Flowable / Self Leveling | 1 minutes | 45 Type A | Download R1-2145 data sheet | Primerless Adhesive;Gray tough, fast cure adhesive with 1 hour pot life. Will RTV. 145 psi (1.0 MPa), unprimed and 625 psi (4.3 MPa) primed (CF1-135) lap shear. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R31-2186 | Platinum | Flowable / Self Leveling | 15 minutes | 20 Type A | Download R31-2186 data sheet | RTV Primerless Adhesive;Very fast cure. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R32-2186 | Platinum | Flowable / Self Leveling | 15 hours | 15 Type A | Download R32-2186 data sheet | Primerless Adhesive;Heat Cure. Flow 0.5 per 5 minutes. Requires heat to cure. 150 psi (1.0 MPa), unprimed lap shear. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R33-2186 | Platinum | Flowable / Self Leveling | 2 hours | 20 Type A | Download R33-2186 data sheet | RTV Primerless Adhesive;Fast cure. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R33-2186-1 | N/A | Flowable / Self Leveling | 2 hours | 20 Type A | Download R33-2186-1 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R4-1140 | Acetoxy | Flowable / Self Leveling | N/A | 25 Type A | Download R4-1140 data sheet | Primerless Adhesive;Self leveling. General all purpose adhesive for a variety of substrates. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SCV-2585 | Platinum | Flowable / Self Leveling | 1 hours | 35 Type A | Download SCV-2585 data sheet | Ultra low Outgassing. Recommended for broad temperature applications. 475 psi (3.3 MPa), primed (CF1-135) lap shear. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SCV-2586 | Platinum | Thixotropic / non-slump | 4 hours | 45 Type A | Download SCV-2586 data sheet | Fast Cure, Low Density, Primed Lap Shear 175psi | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SCV-2590 | Platinum | Flowable / Self Leveling | N/A | 45 Type A | Download SCV-2590 data sheet | Pourable | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SCV-2590-2 | Platinum | Flowable / Self Leveling | 4.5 hours | 50 Type A | Download SCV-2590-2 data sheet | Low Viscosity, Fast Cure | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SCV1-2590 | Platinum | Flowable / Self Leveling | 4 hours | 50 Type A | Download SCV1-2590 data sheet | *4) Primed Lap Shear 175 psi (1.2 MPa) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SCV2-2590 | Platinum | Flowable / Self Leveling | N/A | 45 Type A | Download SCV2-2590 data sheet | Low / High Temperature, *4) Primed Lap Shear 250 psi (1.7 MPa) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
Coatings/Dispersions
Cure System, Durometer,Product | Cure System | Durometer | Data Sheet | Comment | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Acetoxy | 40 Type A | Download data sheet | 240*3) psi (1.7Mpa) Lap Shear at 7 D @ R.T., H / Fed. Color Standard 36118 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-1144-0 | Oxime | N/A | Download CV-1144-0 data sheet | 60% Solids, Atomic Oxygen Protective Overcoat | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-1146-2 | Oxime | N/A | Download CV-1146-2 data sheet | 72% Solids | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-1152 | Oxime | N/A | Download CV-1152 data sheet | Soft, very low modulus coating | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV1-1144-0 | Oxime | N/A | Download CV1-1144-0 data sheet | 50% Solids | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV1-1146-2 | Oxime | N/A | Download CV1-1146-2 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV1-1148 | Oxime | N/A | Download CV1-1148 data sheet | 40% Solids | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV2-1147 | Oxime | N/A | Download CV2-1147 data sheet | 60% Solids, Non-blocking Overcoat | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV3-1144-1 | Oxime | N/A | Download CV3-1144-1 data sheet | 60% Solids | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-2014 | Platinum | N/A | Download MED-2014 data sheet | One Part, Requires heat to cure | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-2214 | Platinum | 35 Type A | Download MED-2214 data sheet | One Part, Requires heat to cure | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4159 | Amine | N/A | Download MED-4159 data sheet | Low Crosslink Density, Amino Functional Lubricious Silicone Coating | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4162 | Non-Curing | N/A | Download MED-4162 data sheet | Lubricious Coating | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6400 | Platinum | 30 Type A | Download MED-6400 data sheet | 1.43 R.I. in Xylene | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6600 | Platinum | 25 Type A | Download MED-6600 data sheet | 1.46 R.I. in Xylene | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6601 | N/A | 24 Type A | Download MED-6601 data sheet | 100% solids | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6605 | Acetoxy | 25 Type A | Download MED-6605 data sheet | One Part, in Xylene | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6615 | Platinum | 72 Type 00 | Download MED-6615 data sheet | Silicone adhesive dispersed in heptane, cures rapidly with heat | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6640 | Platinum | 40 Type A | Download MED-6640 data sheet | Ultra High Tear, in Xylene | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6655 | Acetoxy | 35 Type A | Download MED-6655 data sheet | 100M% Fluorosilicone | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6670 | Platinum | N/A | Download MED-6670 data sheet | Low Coefficient of Friction Coating | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6671 | Acetoxy | N/A | Download MED-6671 data sheet | RTV, Low Coefficient of Friction Coating | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED1-4161 | Condensation | 25 Type 00 | Download MED1-4161 data sheet | 25 '00' durometer lubricious coating | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED10-4161 | Amine | 25 Type 00 | Download MED10-4161 data sheet | Amino Functional Lubricious Silicone Coating | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED10-6400 | Platinum | 30 Type A | Download MED10-6400 data sheet | 1.43 R.I. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED10-6600 | Platinum | 25 Type A | Download MED10-6600 data sheet | 1.46 R.I. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED10-6605 | Acetoxy | 25 Type A | Download MED10-6605 data sheet | One Part | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED10-6607 | Oxime | 40 Type A | Download MED10-6607 data sheet | One Part | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED10-6615 | Platinum | 72 Type 00 | Download MED10-6615 data sheet | Silicone adhesive dispersed in heptane, cures rapidly with heat | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED10-6640 | Platinum | 40 Type A | Download MED10-6640 data sheet | Ultra High Tear | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED10-6655 | Acetoxy | 35 Type A | Download MED10-6655 data sheet | 100M% Fluorosilicone | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED10-6670 | Platinum | N/A | Download MED10-6670 data sheet | Low Coefficient of Friction Coating | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED10-6671 | Acetoxy | N/A | Download MED10-6671 data sheet | RTV, Low Coefficient of Friction Coating | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED11-6604 | Acetoxy | 15 Type A | Download MED11-6604 data sheet | One Part, Smooth Finish | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED16-6606 | Acetoxy | 20 Type A | Download MED16-6606 data sheet | Silicone adhesive dispersed in heptane | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED6-6606 | Acetoxy | 20 Type A | Download MED6-6606 data sheet | Silicone adhesive dispersed in heptane | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-1008-0 | Oxime | 23 Type A | Download R-1008-0 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-1009 | Oxime | 45 Type A | Download R-1009 data sheet | High strength coating recommend for coating PCBs and other electronic assemblies. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-1077 | N/A | 40 Type A | Download R-1077 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-1082 | Acetoxy | 25 Type A | Download R-1082 data sheet | Great adhesion to a variety of substrates. Not recommended where corrosion is of concern. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-1099 | Condensation | 40 Type A | Download R-1099 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-1182 | Acetoxy | N/A | Download R-1182 data sheet | One Part and cures at room temperature. Acetic acid cure by-product therefore not recommended where corrosion is of concern. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2180 | Platinum | 40 Type A | Download R-2180 data sheet | High strength coating and primer is recommended to increase adhesion. Requires heat to cure. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2180-2 | Platinum | 45 Type A | Download R-2180-2 data sheet | Cure: 30 M / 25 C : 45 M / 75 C : 135 M / 150 C, 20% Solids | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2182 | Platinum | N/A | Download R-2182 data sheet | Requires minimum heat to cure. Acetic acid cure by-product therefore not recommended where corrosion is of concern. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-3930 | Acetoxy | 30 Type A | Download R-3930 data sheet | 100%M Fluoro, fuel and hydrocarbon solvent resistant high strength coating. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-3975 | Acetoxy | 25 Type A | Download R-3975 data sheet | Dispersion Coating, 60% Solids | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R1-3930-1 | Acetoxy | 37 Type A | Download R1-3930-1 data sheet | Slight addition of heat will accelerate cure | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R3-1075 | Oxime | 40 Type A | Download R3-1075 data sheet | Dispersion Coating / Conformal, 60% Solids | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R4-3930-11 | Acetoxy | 45 Type A | Download R4-3930-11 data sheet | For protecting or bonding hardware components exposed to gasoline or aviation fuels | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
Elastomer Gels
INCI Name, D4 Content, Viscosity, Appearance,Product | INCI Name | D4 Content | Viscosity | Appearance | Data Sheet | Comment | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CXG-1104 | Dimethicone (and) Dimethicone/Vinyl Dimethicone Crosspolymer | <0.05 % | 635,000 cP | Clear | Download CXG-1104 data sheet | 12% Solids Elastomer Gel Dispersed in Low Viscosity Dimethicone Fluid | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
Electrically Conductive/Static Dissipative
Volume Resistivity, Viscosity, Cure System, Durometer, Tensile,Product | Volume Resistivity | Viscosity | Cure System | Durometer | Tensile | Data Sheet | Comment | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CV-1500 | 3 ohmcm | N/A | Oxime | 80 Type A | 650 psi | Download CV-1500 data sheet | Carbon filled, 3 ohmcm, versatile adhesive. Tack Free in 10 minutes. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2640 | 2.5 ohmcm | N/A | Platinum | 75 Type A | 475 psi | Download CV-2640 data sheet | 2.5 ohmcm, *4) Primed Lap Shear 250 psi (1.7 MPa), Carbon Fiber Filled | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2644 | 0.01 ohmcm | 224,500 cP | Platinum | 85 Type A | 525 psi | Download CV-2644 data sheet | 0.005 ohmcm, *22) 1.2 W/mK | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2646 | 0.01 ohmcm | N/A | Alkoxy | 80 Type A | 400 psi | Download CV-2646 data sheet | Electrically Conductive, 0.007 ohmcm | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV1-2640 | 25 ohmcm | N/A | Platinum | 40 Type A | 525 psi | Download CV1-2640 data sheet | Carbon filled. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV1-2646 | 0.01 ohmcm | N/A | Alkoxy | 80 Type A | 400 psi | Download CV1-2646 data sheet | 0.005 ohmcm | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV2-1148 | 9,000,000,000 ohmcm | N/A | Oxime | 45 Type A | N/A | Download CV2-1148 data sheet | 100% Solids | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV2-2640 | 1 ohmcm | 675,000 cP | Platinum | 30 Type A | 500 psi | Download CV2-2640 data sheet | Carbon Black Filled | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV2-2644 | 0 ohmcm | N/A | Platinum | 85 Type A | 500 psi | Download CV2-2644 data sheet | 0.004 ohmcm | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV3-2640 | 2,200,000 ohmcm | 14,100 cP | Platinum | 25 Type A | 70 psi | Download CV3-2640 data sheet | Carbon filled. Designed for broad operating temperature. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-1505 | 8 ohmcm | N/A | Oxime | 75 Type A | 525 psi | Download R-1505 data sheet | 8 ohmcm. Static Dissipation | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2631 | 70 ohmcm | N/A | Platinum | 45 Type A | 615 psi | Download R-2631 data sheet | Carbon filled. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2637 | 0.01 ohmcm | N/A | Platinum | 60 Type A | 210 psi | Download R-2637 data sheet | Electrically Conductive RTV Silicone. Mixes to smooth paste, can be screen printed. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SCV-2596 | 2.5 ohmcm | N/A | Platinum | 75 Type A | 475 psi | Download SCV-2596 data sheet | Electrically Conductive, 2.5 ohmcm, Carbon Fiber Filled | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SCV1-2596 | 0.01 ohmcm | N/A | Platinum | 85 Type A | 450 psi | Download SCV1-2596 data sheet | Electricaly Conductive, 0.005 ohmcm ,*21, 22) 1.2 W /mK | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
Encapsulation Technology
INCI Name, D4 Content, Viscosity, Appearance,Product | INCI Name | D4 Content | Viscosity | Appearance | Data Sheet | Comment | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CES-1104 | Dimethicone and Aqua and Glycerin and Pentylene Glycol and Dimethicone/Vinyl Dimethicone Crosspolymer and Phenoxyethanol and Carbomer and Amodimethicone and Sodium Hydroxide and Disodium EDTA | <0.05 % | 60,000 cP | Translucent | Download CES-1104 data sheet | Encapsulated Silicone with 12% CXG-1104 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
Fluids
Substrate, Volatile Content, Viscosity,Product | Substrate | Volatile Content | Viscosity | Data Sheet | Comment | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
FS-3602 | Silicone | 0.1 % | N/A | Download FS-3602 data sheet | N/A | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-360 | Glass / Metal / Plastic | 0.1 % | 100 cP / 350 cP / 1,000 cP / 12,500 cP | Download MED-360 data sheet | Dimethyl Polymer | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-361 | Glass / Metal / Plastic | 0.1 % | 100 cP / 350 cP / 1,000 cP / 12,500 cP | Download MED-361 data sheet | Dimethyl Polymer | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-366 | N/A | 0.1 % | 1,000 cP | Download MED-366 data sheet | Highly purified Dimethyl Polymer | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-367 | N/A | 0.1 % | 5,000 cP | Download MED-367 data sheet | Highly purified Dimethyl Polymer | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-400 | Silicone | 0.3 % | 350 cP / 1,000 cP / 12,500 cP | Download MED-400 data sheet | Fluorosilicone Polymer | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-420 | Silicone | 0.1 % | 350 cP / 1,000 cP / 12,500 cP | Download MED-420 data sheet | Methyl Fluoro Copolymer (Low Fluoro Content) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-460 | Silicone | 0.1 % | 350 cP / 1,000 cP / 12,500 cP | Download MED-460 data sheet | Methyl Fluoro Copolymer (High Fluoro Content) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
PLY-7602 | N/A | N/A | N/A | Download PLY-7602 data sheet | A processing aid or plasticizer for elastomeric systems | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
PLY-7700 | N/A | 48 % | 6 cSt | Download PLY-7700 data sheet | Low viscosity fluid | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
S-7200 | Glass / Metal / Plastic | 2 % | 7,000,000 cP | Download S-7200 data sheet | 100% Dimethylsiloxane non-reactive polymer. Custom viscosities available. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
S-7400 | Glass / Metal / Plastic | 4 % | 60,000 cP | Download S-7400 data sheet | Recommend for extreme temperature (-100°C to 250°C). Higher and custom viscosities available. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
Foams
Foam Density, Viscosity, Cure System, Application Life, Appearance,Product | Foam Density | Viscosity | Cure System | Application Life | Appearance | Data Sheet | Comment | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CF1-3710-2 | 50 lbs/ft3 | N/A | Platinum | N/A | Gray | Download CF1-3710-2 data sheet | Cure in place fuel resistant foam for short term fuel exposure, application life 10 minutes | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CF3-2350 | 25 lbs/ft³ | 90,000 cP | Platinum | N/A | Gray | Download CF3-2350 data sheet | Useful where stability at higher and lower temperatures is required. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2391 | 20 lbs/ft3 | 3,000 cP | Platinum | 6 minutes | White | Download CV-2391 data sheet | Cure in place versatile low outgassing foam, application life 6 minutes | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-2310 | N/A | 40,000 cP | Platinum | 2 minutes | White | Download MED-2310 data sheet | Low Density | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED10-2310 | N/A | 40,000 cP | Platinum | 2 minutes | White | Download MED10-2310 data sheet | Low Density | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2360 | 12 lbs/ft3 | 40,000 cP | Platinum | 2 minutes | White | Download R-2360 data sheet | Flexible cure in place low density foam. Application life 2 minutes. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2370 | 10 lbs/ft3 | 4,700 cP | Tin | 28 minutes | Tan | Download R-2370 data sheet | Low Density / Soft, 10 lb/ft^3 (0.16 g/cm^3) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SFM5-2350 | 25 lbs/ft3 | 52,500 cP | Platinum | N/A | Black | Download SFM5-2350 data sheet | Cure in place versatile tough foam. Application life 6 minutes. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
Gels
Viscosity, Penetration, Work Time, Mix Ratio,Product | Viscosity | Penetration | Work Time | Mix Ratio | Data Sheet | Comment | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CV-8151 | 2,500 cP | 4 mm | 30 hours | 1:1 | Download CV-8151 data sheet | Low Viscosity, Penetration 4.0 mm | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-8251 | 1,800 cP | 3 mm | 24 hours | 1:1 | Download CV-8251 data sheet | Low/High Temperature, Penetration 3 mm | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
EPM-2480 | 2,500 cP | 4 mm | 30 hours | 1:1 | Download EPM-2480 data sheet | Soft High Tack Gel | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
EPM-2482 | 1,800 cP | 3 mm | N/A | 1:1 | Download EPM-2482 data sheet | Soft High Tack Gel;For applications requiring low volatility. Useful for potting intricate assemblies due to a low viscosity. As a low modulus embedding or potting compound for environmental protection of electronic assemblies and components | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
GEL-3500 | 11,250 cP | N/A | 12 hours | 1:1 | Download GEL-3500 data sheet | Firm Fluoro Gel;Recommended for short term exposure to fuels and solvent | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
GEL-8100 | 535 cP | N/A | N/A | 1:1 | Download GEL-8100 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
GEL-8111 | 535 cP | N/A | N/A | 1:1 | Download GEL-8111 data sheet | For applications requiring a low viscosity and low volatility soft gel | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
GEL-8127 | 530 cP | 8 mm | N/A | N/A | Download GEL-8127 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
GEL-8136 | 450 cP | 14 mm | N/A | N/A | Download GEL-8136 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
GEL-8150 | 500 cP | 5 mm | 4 hours | 1:1 | Download GEL-8150 data sheet | Soft High Tack Gel | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
GEL-8170 | 600 cP | 9 mm | N/A | 1:1 | Download GEL-8170 data sheet | Very Soft High Tack Gel;Max viscosity after 6 hrs is 2,000 cP | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
GEL-8250 | 650 cP | 5 mm | 2 hours | 1:1 | Download GEL-8250 data sheet | Soft Med Tack Gel;For applications requiring a broader operating temperature range. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
GEL1-8155 | 14,500 cP | 0.4 mm | N/A | 1:1 | Download GEL1-8155 data sheet | Soft Low Tack Gel;Viscosity increases ~ 1.5X in 24 hrs (¼” diameter foot, 1.5 seconds) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
GEL8-8150 | 500 cP | 4 mm | 1.5 hours | 1:1 | Download GEL8-8150 data sheet | Soft High Tack Gel | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
LS-3238 | 1,500 cP | N/A | 11 hours | 1:1 | Download LS-3238 data sheet | Resistant to hydrocarbon solvents. Index matches Magnesium Fluoride. Potlife based on 3,000 cPs maximum mixed viscosity after 6hrs | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
LS-3441 | 14,500 cP | 0.4 mm | N/A | 1:1 | Download LS-3441 data sheet | Soft Low Tack Gel; Tough gel and 24 hr minimum potlife based on 2X initial viscosity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
LS-3443 | 650 cP | 5 mm | N/A | 1:1 | Download LS-3443 data sheet | Soft High Tack Gel;Recommended as an optically robust alter-te to index match to 1.46 substrates;will LTV , 2hr minimum potlife based on 2X initial viscosity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
LS1-3252 | 360 cP | N/A | N/A | N/A | Download LS1-3252 data sheet | 1.52 Refractive Index firm gel for encapsulating LEDs, and to reduce lightloss for other optical applications. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
LS1-3443 | 2,500 cP | 6 mm | N/A | N/A | Download LS1-3443 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
LS3-3354 | 5,200 cP | N/A | 80 minutes | 1:1 | Download LS3-3354 data sheet | Soft Gel;Excellent for phosphor coatings. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6311 | 500 cP | 2.6 mm | N/A | 1:1 | Download MED-6311 data sheet | Low penetration, high strength gel | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED1-6300 | 1,000 cP | 5 mm | N/A | 1:1 | Download MED1-6300 data sheet | Medium penetration, soft responsive gel | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED2-6300 | 1,000 cP | 10 mm | N/A | 1:1 | Download MED2-6300 data sheet | Low penetration, firm cohesive gel | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED3-6300 | 1,000 cP | 15 mm | N/A | 3:1 | Download MED3-6300 data sheet | High penetration, soft responsive gel | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED3-6311 | 500 cP | 2.6 mm | N/A | 3:1 | Download MED3-6311 data sheet | High Purity, Low Penetration, High Strength Gel | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
Greases
Substrate, Volatile Content, Viscosity,Product | Substrate | Volatile Content | Viscosity | Data Sheet | Comment | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CV-9042 | Glass / Metal / Plastic | N/A | N/A | Download CV-9042 data sheet | Thermally Conductive | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-9052 | Glass / Metal / Plastic | N/A | N/A | Download CV-9052 data sheet | Volume Resistivity 1x1015 ohmcm, Thermally conductive | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-9341 | Glass / Metal / Plastic | N/A | N/A | Download CV-9341 data sheet | Thermally Conductive | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
G-9010 | Glass / Metal / Plastic | 0.34 % | 1,100,000 cP | Download G-9010 data sheet | A stiff methyl silicone/Teflon grease for high vacuum and pressure systems. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
G-9020 | Glass / Metal / Plastic | 0.3 % | 310,000 cP | Download G-9020 data sheet | A medium consistency methyl silicone grease for high vacuum and pressure systems. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
G-9030 | Glass / Metal / Plastic | 0.3 % | 980,100 cP | Download G-9030 data sheet | A stiff methyl grease used lubricating . 0.3% Weight loss. For high vacuum and pressure systems. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
LS-1246 | Glass / Metal / Plastic | N/A | N/A | Download LS-1246 data sheet | N/A | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6731 | Silicone | 0.6 % | 2,000,000 cP | Download MED-6731 data sheet | High Consistency Methyl Fluoro Grease | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-9011 | Glass / Metal / Plastic | 0.3 % | 95,000 cP | Download MED-9011 data sheet | Low Consistency Grease | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-9021 | Glass / Metal / Plastic | 0.5 % | 300,000 cP | Download MED-9021 data sheet | Medium Consistency Grease | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-9031 | Glass / Metal / Plastic | 0.3 % | 900,000 cP | Download MED-9031 data sheet | High Consistency Grease | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
High Consistency Rubbers
Durometer, Tensile, Elongation, Tear,Product | Durometer | Tensile | Elongation | Tear | Data Sheet | Comment | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
MED-2174 | 50 Type A | 1,300 psi | 750 % | 225 ppi | Download MED-2174 data sheet | Uncatalyzed | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4014 | 15 Type A | 700 psi | 1,350 % | 155 ppi | Download MED-4014 data sheet | Low modulus | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4019 | 55 Type A | 1,290 psi | 645 % | 205 ppi | Download MED-4019 data sheet | Excellent adhesion and fatigue resistance | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4020 | 25 Type A | 1,400 psi | 1,250 % | 190 ppi | Download MED-4020 data sheet | Low modulus with high tear | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4025 | 30 Type A | 1,300 psi | 900 % | 130 ppi | Download MED-4025 data sheet | Low tension set | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4027 | 30 Type A | 2,300 psi | 1,050 % | 235 ppi | Download MED-4027 data sheet | Ultra-high tensile strength | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4032 | 30 Type A | 1,200 psi | 1,100 % | 160 ppi | Download MED-4032 data sheet | Can be Cured via Platinum or Peroxide | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4035 | 35 Type A | 1,565 psi | 1,055 % | 195 ppi | Download MED-4035 data sheet | High Tear Strength | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4042 | 40 Type A | 1,500 psi | 1000 % | 160 ppi | Download MED-4042 data sheet | Can be Cured via Platinum or Peroxide | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4050 | 50 Type A | 1,500 psi | 985 % | 250 ppi | Download MED-4050 data sheet | High Tear Strength | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4052 | 50 Type A | 1,445 psi | 1100 % | 230 ppi | Download MED-4052 data sheet | Can be Cured via Platinum or Peroxide | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4055 | 55 Type A | 1,700 psi | 900 % | 300 ppi | Download MED-4055 data sheet | Ultra-high tear strength | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4062 | 60 Type A | 1,400 psi | 1000 % | 250 ppi | Download MED-4062 data sheet | Can be Cured via Platinum or Peroxide | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4065 | 65 Type A | 1,250 psi | 1000 % | 250 ppi | Download MED-4065 data sheet | High Tear Strength | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4070 | 70 Type A | 1,430 psi | 750 % | 300 ppi | Download MED-4070 data sheet | Abrasion / Fatigue Resistant | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4072 | 70 Type A | 1,100 psi | 875 % | 240 ppi | Download MED-4072 data sheet | Can be Cured via Platinum or Peroxide | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4080 | 80 Type A | 1,150 psi | 700 % | 225 ppi | Download MED-4080 data sheet | High Tear Strength | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4082 | 80 Type A | 1,150 psi | 900 % | 200 ppi | Download MED-4082 data sheet | Can be Cured via Platinum or Peroxide | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4120 | 30 Type A | 1,300 psi | 950 % | 140 ppi | Download MED-4120 data sheet | Uncatalyzed | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4128 | 25 Type A | 1,010 psi | 800 % | 70 ppi | Download MED-4128 data sheet | Low Tension Set, Uncatalyzed | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4135 | 35 Type A | 1,250 psi | 800 % | 110 ppi | Download MED-4135 data sheet | Uncatalyzed | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4150 | 50 Type A | 1,400 psi | 750 % | 190 ppi | Download MED-4150 data sheet | Uncatalyzed | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4174 | 50 Type A | 1,200 psi | 775 % | 225 ppi | Download MED-4174 data sheet | Uncatalyzed | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4520 | 28 Type A | 1,320 psi | 950 % | 135 ppi | Download MED-4520 data sheet | Uncatalyzed | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4550 | 50 Type A | 1,400 psi | 750 % | 195 ppi | Download MED-4550 data sheet | Uncatalyzed | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4565 | 65 Type A | 1,250 psi | 700 % | 200 ppi | Download MED-4565 data sheet | Uncatalyzed | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4714 | 15 Type A | 715 psi | 1385 % | 155 ppi | Download MED-4714 data sheet | Low Durometer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4719 | 55 Type A | 1,320 psi | 665 % | 220 ppi | Download MED-4719 data sheet | Excellent adhesion and fatigue resistance | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4720 | 25 Type A | 1,390 psi | 1240 % | 190 ppi | Download MED-4720 data sheet | High Tear | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4725 | 30 Type A | 1,395 psi | 890 % | 135 ppi | Download MED-4725 data sheet | Low Tension Set | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4727 | 30 Type A | 2,300 psi | 1050 % | 235 ppi | Download MED-4727 data sheet | High Strength, Low Modulus | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4735 | 35 Type A | 1,535 psi | 1055 % | 195 ppi | Download MED-4735 data sheet | High Tear | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4750 | 50 Type A | 1,545 psi | 1050 % | 270 ppi | Download MED-4750 data sheet | High Tear | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4755 | 55 Type A | 1,640 psi | 875 % | 310 ppi | Download MED-4755 data sheet | Ultra-high Tear | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4765 | 65 Type A | 1,240 psi | 1005 % | 265 ppi | Download MED-4765 data sheet | High Tear | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4770 | 70 Type A | 1,415 psi | 760 % | 300 ppi | Download MED-4770 data sheet | Abrasion / Fatigue Resistant | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4780 | 80 Type A | 1,155 psi | 740 % | 225 ppi | Download MED-4780 data sheet | High Tear | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED4-4115 | 50 Type A | 1,600 psi | 500 % | 100 ppi | Download MED4-4115 data sheet | Non-Vinyl Specific Peroxide Precatalyzed | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED4-4116 | 70 Type A | 1,300 psi | 450 % | 125 ppi | Download MED4-4116 data sheet | Non-Vinyl Specific Peroxide Precatalyzed | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED4-4515 | 50 Type A | 1,650 psi | 500 % | 100 ppi | Download MED4-4515 data sheet | Non-Vinyl Specific Peroxide Precatalyzed | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED4-4516 | 70 Type A | 1,300 psi | 450 % | 130 ppi | Download MED4-4516 data sheet | Non-Vinyl Specific Peroxide Precatalyzed | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SIL2-5020 | 21 Type A | 1,620 psi | 1185 % | 195 ppi | Download SIL2-5020 data sheet | Designed For High Volume Extrusion Or Molding | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SIL2-5030 | 31 Type A | 1,690 psi | 1140 % | 215 ppi | Download SIL2-5030 data sheet | Designed for High Volume Extrusion or Molding | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SIL2-5040 | 39 Type A | 1,510 psi | 1115 % | 220 ppi | Download SIL2-5040 data sheet | Designed for High Volume Extrusion or Molding | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SIL2-5050 | 50 Type A | 1,350 psi | 940 % | 245 ppi | Download SIL2-5050 data sheet | Designed for High Volume Extrusion or Molding | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SIL2-5060 | 60 Type A | 1,300 psi | 920 % | 255 ppi | Download SIL2-5060 data sheet | Designed for High Volume Extrusion or Molding | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SIL2-5070 | 68 Type A | 1,330 psi | 855 % | 260 ppi | Download SIL2-5070 data sheet | Designed for High Volume Extrusion or Molding | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SIL2-5080 | 80 Type A | 1,140 psi | 615 % | 210 ppi | Download SIL2-5080 data sheet | Designed for High Volume Extrusion or Molding | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
Inks
Appearance, Mix Ratio, Non Volatile Content, Viscosity,Product | Appearance | Mix Ratio | Non Volatile Content | Viscosity | Data Sheet | Comment | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
MED-6608-1 | White | 1 Part | 70 % | 1,150 cP | Download MED-6608-1 data sheet | RTV Ink in White | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6608-2 | Black | 1 Part | 70 % | 1,150 cP | Download MED-6608-2 data sheet | RTV Ink in Black | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6613-1 | White | 1:1 | 65 % | N/A | Download MED-6613-1 data sheet | Heat Curable Ink in White | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6613-2 | Black | 1:1 | 60 % | 800 cP | Download MED-6613-2 data sheet | Heat Curable Ink in Black | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6613-7 | Dark Blue | 1:1 | 65 % | N/A | Download MED-6613-7 data sheet | Heat Curable Ink in Dark Blue | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED8-6608-2 | Black | 1 PART | 70 % | 800 cP | Download MED8-6608-2 data sheet | RTV Ink in Black, Increased Hiding Power | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-1008-1 | White | 1 PART | 70 % | 1,150 cP | Download R-1008-1 data sheet | Available in clear (R-1008-0), white (R-1008-1), black (R-1008-2), red (R-1008-3), orange (R-1008-4), yellow (R-1008-5), green (R-1008-6), blue (R-1008-7), and violet (R-1008-8). | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-1008-2 | Black | 1 PART | 70 % | 1,150 cP | Download R-1008-2 data sheet | Available in clear (R-1008-0), white (R-1008-1), black (R-1008-2), red (R-1008-3), orange (R-1008-4), yellow (R-1008-5), green (R-1008-6), blue (R-1008-7), and violet (R-1008-8). | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-1008-3 | Red | 1 PART | 70 % | 1,150 cP | Download R-1008-3 data sheet | Available in clear (R-1008-0), white (R-1008-1), black (R-1008-2), red (R-1008-3), orange (R-1008-4), yellow (R-1008-5), green (R-1008-6), blue (R-1008-7), and violet (R-1008-8). | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-1008-4 | Orange | 1 PART | 70 % | 1,150 cP | Download R-1008-4 data sheet | Available in clear (R-1008-0), white (R-1008-1), black (R-1008-2), red (R-1008-3), orange (R-1008-4), yellow (R-1008-5), green (R-1008-6), blue (R-1008-7), and violet (R-1008-8). | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-1008-5 | Yellow | 1 PART | 70 % | 1,150 cP | Download R-1008-5 data sheet | Available in clear (R-1008-0), white (R-1008-1), black (R-1008-2), red (R-1008-3), orange (R-1008-4), yellow (R-1008-5), green (R-1008-6), blue (R-1008-7), and violet (R-1008-8). | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-1008-6 | Green | 1 PART | 70 % | 1,150 cP | Download R-1008-6 data sheet | Available in clear (R-1008-0), white (R-1008-1), black (R-1008-2), red (R-1008-3), orange (R-1008-4), yellow (R-1008-5), green (R-1008-6), blue (R-1008-7), and violet (R-1008-8). | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-1008-7 | Blue | 1 PART | 70 % | 1,150 cP | Download R-1008-7 data sheet | Available in clear (R-1008-0), white (R-1008-1), black (R-1008-2), red (R-1008-3), orange (R-1008-4), yellow (R-1008-5), green (R-1008-6), blue (R-1008-7), and violet (R-1008-8). | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-1008-8 | Violet | 1 PART | 70 % | 1,150 cP | Download R-1008-8 data sheet | Available in clear (R-1008-0), white (R-1008-1), black (R-1008-2), red (R-1008-3), orange (R-1008-4), yellow (R-1008-5), green (R-1008-6), blue (R-1008-7), and violet (R-1008-8). | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2100-2 | Black | 1:1 | 60 % | 900 cP | Download R-2100-2 data sheet | Available in white (R-2100-1), black (R-2100-2), red (R-2100-3), and green (R-2100-6). Dispersed in xylene. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
Liquid Silicone Rubbers
Durometer, Tensile, Elongation, Tear,Product | Durometer | Tensile | Elongation | Tear | Data Sheet | Comment | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
FS-3511 | 40 Type A | 1,150 psi | 335 % | 60 ppi | Download FS-3511 data sheet | LSR Fluoro Elastomer;For long term fuel or solvent exposure. Can be liquid injection molded and requires heat to cure. Recommend SP-120 or SP-201 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4801 | 38 Type 00 | 290 psi | 1140 % | 55 ppi | Download MED-4801 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4805 | 7 Type A | 485 psi | 1080 % | 70 ppi | Download MED-4805 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4810 | 11 Type A | 700 psi | 1075 % | 65 ppi | Download MED-4810 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4820 | 20 Type A | 910 psi | 875 % | 125 ppi | Download MED-4820 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4830 | 30 Type A | 1,350 psi | 750 % | 150 ppi | Download MED-4830 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4840 | 43 Type A | 1,190 | 560 % | 245 ppi | Download MED-4840 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4842 | 44 Type A | 1,070 psi | 400 % | 250 ppi | Download MED-4842 data sheet | Injection Molding Elastomer, Resists Blocking | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4850 | 50 Type A | 1,475 psi | 675 % | 240 ppi | Download MED-4850 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4860 | 60 Type A | 1,350 psi | 530 % | 255 ppi | Download MED-4860 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4870 | 70 Type A | 1,500 psi | 415 % | 230 ppi | Download MED-4870 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4880 | 80 Type A | 1,030 psi | 265 % | 90 ppi | Download MED-4880 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4901 | 40 Type 00 | 285 psi | 1135 % | 55 ppi | Download MED-4901 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4905 | 7 Type A | 350 psi | 1000 % | 70 ppi | Download MED-4905 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4910 | 10 Type A | 450 psi | 1000 % | 65 ppi | Download MED-4910 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4920 | 20 Type A | 750 psi | 700 % | 125 ppi | Download MED-4920 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4930 | 30 Type A | 800 psi | 450 % | 140 ppi | Download MED-4930 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4940 | 40 Type A | 1,190 psi | 560% | 246 ppi | Download MED-4940 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4942 | 40 Type A | 850 psi | 350 % | 130 ppi | Download MED-4942 data sheet | Injection Molding Elastomer, Resists Blocking | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4950 | 50 Type A | 1,445 psi | 700% | 243 ppi | Download MED-4950 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4960 | 60 Type A | 1,300 psi | 525 % | 250 ppi | Download MED-4960 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4970 | 70 Type A | 1,325 psi | 465 % | 240 ppi | Download MED-4970 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4971 | 68 Type A | 875 psi | 230 % | 170 ppi | Download MED-4971 data sheet | High Clarity Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4980 | 80 Type A | 1,000 psi | 250 % | 90 ppi | Download MED-4980 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-5440 | 30 Type A | 1,250 psi | 380 % | 60 ppi | Download MED-5440 data sheet | 100M% Fluorosilicone Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-5820 | 19 Type A | 1,080 psi | 890 % | 90 ppi | Download MED-5820 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-5830 | 30 Type A | 1,340 psi | 830 % | 150 ppi | Download MED-5830 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-5840 | 40 Type A | 1,350 psi | 680 % | 160 ppi | Download MED-5840 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-5850 | 50 Type A | 1,450 psi | 550 % | 190 ppi | Download MED-5850 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-5860 | 60 Type A | 1,300 psi | 425 % | 200 ppi | Download MED-5860 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-5870 | 68 Type A | 1,250 psi | 400 % | 225 ppi | Download MED-5870 data sheet | Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED1-4855 | 55 Type A | 1,100 psi | 475 % | 240 ppi | Download MED1-4855 data sheet | Self-Lubricating Injection Molding Elastomer, High Lubrication | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED1-4955 | 55 Type A | 1,165 psi | 490 % | 250 ppi | Download MED1-4955 data sheet | Self-Lubricating Injection Molding Elastomer, High Lubrication | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED10-5440 | 30 Type A | 1,250 psi | 380 % | 60 ppi | Download MED10-5440 data sheet | 100M% Fluorosilicone Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED2-4955 | 61 Type A | 1,200 psi | 490 % | 250 ppi | Download MED2-4955 data sheet | Self-Lubricating Injection Molding Elastomer, Moderate Lubrication | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED30-4940-1 | 35 Type A | 970 psi | 570 % | 210 ppi | Download MED30-4940-1 data sheet | Self-Lubricating Injection Molding Elastomer, Low Lubrication | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED50-5338 | 30 Type A | 650 psi | 350 % | 37 ppi | Download MED50-5338 data sheet | 50M% Fluorosilicone Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED50-5438 | 31 Type A | 650 psi | 350 % | 36 ppi | Download MED50-5438 data sheet | 50M% Fluorosilicone Injection Molding Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
Low Viscosity Elastomers
Viscosity, Durometer, Work Time, Tensile,Product | Viscosity | Durometer | Work Time | Tensile | Data Sheet | Comment | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CF1-3510 | 70,000 cP | 20 Type A | 7 hours | 185 psi | Download CF1-3510 data sheet | LCE Fluoro Elastomer;100% Fluorosilicone, flowable mixed viscosity, and 7 hr. pot life. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CF19-2186 | 75,000 cP | 25 Type A | 15 minutes | 1,100 psi | Download CF19-2186 data sheet | Dielectric properties are excellent for actuators and other applications where dielectric properties are critical | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CF19-2615 | 1,300 cP | 30 Type A | 4 hours | 120 psi | Download CF19-2615 data sheet | Versatile, clear coating, cure can be heat accelerated. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV4-2500 | 1,500 cP | 28 Type A | N/A | N/A | Download CV4-2500 data sheet | Low Outgassing clear coating, cure can be heat accelerated. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
EPM-2420 | 2,600 cP | 30 Type A | N/A | 425 psi | Download EPM-2420 data sheet | Clear, self leveling and good adhesion to various plastics. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
EPM-2421 | 3,225 cP | 50 Type A | 3 hours | 850 psi | Download EPM-2421 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
LS-6140 | 3,125 cP | 50 Type A | 3 hours | 850 psi | Download LS-6140 data sheet | RTV; Low volatility | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
LS-6941 | 5,300 cP | 50 Type A | 5.5 hours | 1,300 psi | Download LS-6941 data sheet | General use RTV clear adhesive | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
LS-6943 | 5,400 cP | 40 Type A | N/A | 900 psi | Download LS-6943 data sheet | 1.43 Refractive Index at 589nm. For applications requiring a broader operating temperature range. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
LS-6946 | 37,500 cP | 30 Type A | 2 hours | 675 psi | Download LS-6946 data sheet | 1.46 Refractive Index at 589nm and contains fumed silica. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
LS-8941 | 21,500 cP | 80 Type A | 24 hours | 1,250 psi | Download LS-8941 data sheet | Hard and flexible elastomer. Can be casted and liquid injection molded. Requires heat to cure. Recommend SP-120 or SP-201 for overmolding. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
LS1-6140 | 3,200 cP | 50 Type A | N/A | 900 psi | Download LS1-6140 data sheet | Low volatility and requires heat to cure, long pot life for dispensing | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
LS1-6941 | 62,500 cP | 50 Type A | 24 hours | 750 psi | Download LS1-6941 data sheet | Versatile and tough clear elastomer. Requires heat to cure. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
LS2-6140 | 2,740 cP | 50 Type A | N/A | 1,130 psi | Download LS2-6140 data sheet | Low Volatility, 1.41 Refractive Index at 589nm. Long pot life and requires heat to cure (80 C). Primerless adhesion to ceramic, plastics and metals. Tested per UL94 and passes V0. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4011 | N/A | 27 Type A | N/A | 670 psi | Download MED-4011 data sheet | Flowable | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4044 | 73,400 cP | 40 Type A | 5 hours | 875 psi | Download MED-4044 data sheet | Flowable, Medium Durometer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4086 | 6,450 cP | 55 Type 000 | 18 hours | 40 psi | Download MED-4086 data sheet | Ultra Soft Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4211 | 97,800 cP | 25 Type A | N/A | 685 psi | Download MED-4211 data sheet | Flowable | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4244 | 130,000 cP | 40 Type A | 4 hours | 865 psi | Download MED-4244 data sheet | Flowable, Medium Durometer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4286 | 6,350 cP | 55 Type 000 | 17.5 hours | 45 psi | Download MED-4286 data sheet | Ultra Soft Elastomer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-4917 | 11,750 cP | 20 Type A | 31 hours | 500 psi | Download MED-4917 data sheet | Low Viscosity, Long Pot Life | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6010 | 16,000 cP | 50 Type A | 4 hours | 950 psi | Download MED-6010 data sheet | 1.43 R.I., Optically Clear | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6015 | 3,800 cP | 50 Type A | 5.5 hours | 1,200 psi | Download MED-6015 data sheet | 1.41 R.I., Optically Clear, Low Viscosity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6019 | 25,500 cP | 75 Type A | 2.5 hours | 1,300 psi | Download MED-6019 data sheet | Low Viscosity, High Durometer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6020 | 65,000 cP | 40 Type A | 4 hours | 640 psi | Download MED-6020 data sheet | 1.43 R.I., Optically Clear | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6033 | 72,000 cP | 50 Type A | 50 hours | 730 psi | Download MED-6033 data sheet | 1.41 R.I., Optically Clear, Pourable | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6210 | 16,000 cP | 50 Type A | 4 hours | 930 psi | Download MED-6210 data sheet | Clear, 1.43 R.I. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6215 | 3,800 cP | 50 Type A | 5 hours | 1,250 psi | Download MED-6215 data sheet | Clear, 1.41 R.I. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6219 | 25,500 cP | 75 Type A | 2.5 hours | 1,300 psi | Download MED-6219 data sheet | Clear, 1.41 R.I., Low Viscosity, High Durometer | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6233 | 73,500 cP | 50 Type A | 52 hours | 740 psi | Download MED-6233 data sheet | Clear, 1.41 R.I. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6820 | 66,000 cP | 40 Type A | 4.5 hours | 670 psi | Download MED-6820 data sheet | 1.43 R.I., Optically Clear | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED2-4220 | 19,500 cP | 20 Type A | 3 minutes | 585 psi | Download MED2-4220 data sheet | Low Viscosity, Rapid RTV Cure | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED2-4420 | 22,000 cP | 20 Type A | 3 minutes | 585 psi | Download MED2-4420 data sheet | Low Viscosity, Rapid RTV Cure | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED3-4220 | 74,500 cP | 25 Type A | 15 minutes | 1,100 psi | Download MED3-4220 data sheet | Flowable | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED3-4420 | 91,250 cP | 23 Type A | 15 minutes | 1,000 psi | Download MED3-4420 data sheet | Flowable | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED4-4220 | 20,500 cP | 17 Type A | 27 minutes | 660 psi | Download MED4-4220 data sheet | Low Viscosity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED4-4420 | 24,000 cP | 17 Type A | 35 minutes | 700 psi | Download MED4-4420 data sheet | RTV | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2188 | 11,300 cP | 20 Type A | 8 hours | 475 psi | Download R-2188 data sheet | Excellent dielectric properties, flexible cure with 14 hr pot life, tested per UL 94 and passed V1. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2613 | 4,000 cP | 45 Type A | 2 hours | 1,140 psi | Download R-2613 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2615 | 5,300 cP | 50 Type A | 4 hours | 1,300 psi | Download R-2615 data sheet | Self leveling, low temperature cure versatile encapsulant | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2615-3 | N/A | 50 Type A | 2 hours | 1,000 psi | Download R-2615-3 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2655 | 5,400 cP | 40 Type A | N/A | 900 psi | Download R-2655 data sheet | For applications requiring a broader operating temperature range | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R2-6755 | 45,000 cP | 20 Type A | N/A | 550 psi | Download R2-6755 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R21-2615 | 25,000 cP | 75 Type A | 2 hours | 1,200 psi | Download R21-2615 data sheet | Hard clear elastomer with 2 hr pot life and LTV cure | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
Pressure Sensitive Adhesives
Viscosity, Non Volatile Content,Product | Viscosity | Non Volatile Content | Data Sheet | Comment | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CV3-1161 | 1,300 cP | 40 % | Download CV3-1161 data sheet | Clear silicone PSA for adhering and bonding silicone elastomers, metals, paper, glass, fabric, organic rubbers, some synthetics. Excellent for tapes on films such as polyester or kapton.Rlease force of PSA: 1.5 ppi. Same as CV-1161 with peroxide catalyst to reduce flow at higher temperatures. Recommended where low VOCs are of concern. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-1353 | 3,500 cP | 68 % | Download MED-1353 data sheet | High cohesive strength | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-1356 | 250 cP | 50 % | Download MED-1356 data sheet | LOW MODULUS, HIGH TACK | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED1-1356 | 245 cP | 50 % | Download MED1-1356 data sheet | LOW MODULUS, HIGH TACK | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
PSA-1170 | 250 cP | 50 % | Download PSA-1170 data sheet | Use for tapes and fasteners for temporarily adhering various substrates including metals, glass, silicone, and organic rubbers. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
PSA-1180 | 3,300 cP | 70 % | Download PSA-1180 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
PSA-1270 | 2,100 cP | 50 % | Download PSA-1270 data sheet | N/A | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
Primers
Non Volatile Content, Solvent,Product | Non Volatile Content | Solvent | Data Sheet | Comment | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CF1-135 | 4.5 % | Naptha | Download CF1-135 data sheet | Use with addition cure silicones. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CF1-141 | 6 % | IPA | Download CF1-141 data sheet | Addition Cure Systems, IPA Based, 6% Solids | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CF2-135 | 4.5 % | Naptha | Download CF2-135 data sheet | Use with addition cure silicones. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CF6-135 | 9 % | Naptha | Download CF6-135 data sheet | Recommended where platinum inhibition is of concern. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-160 | 4 % | Naptha | Download MED-160 data sheet | General all-purpose primer. Use with platinum or moisture cure silicones. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-162 | 15 % | Naptha | Download MED-162 data sheet | Increased adhesion to polycarbonate (PC), polyurethane (PU) and polyetheretherketone (PEEK). | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-163 | 15 % | Naptha | Download MED-163 data sheet | Improved adhesion to polyimide (PI). Recommended where platinum inhibition is of concern. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-164 | 10 % | Naptha | Download MED-164 data sheet | Increased adhesion to various substrates. Designed for use with moisture cure systems. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-165 | 4.8 % | Naptha | Download MED-165 data sheet | Increased adhesion to various metals. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-166 | 6.5 % | IPA | Download MED-166 data sheet | Dispersed in Isopropyl Alcohol, compatible with acrylics. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED1-161 | 4.5 % | Naptha | Download MED1-161 data sheet | Increased adhesion to aluminum. Use with platinum cure silicones. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED2-161 | 4.5 % | Naptha | Download MED2-161 data sheet | Increased adhesion to most metals. Use with platinum cure systems. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED6-161 | 8.7 % | Naptha | Download MED6-161 data sheet | Increased adhesion to titanium, polysulfone (PSU), polycarbonate (PC), polyurethane (PU). Recommended where platinum inhibition is of concern. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SP-120 | 4 % | Naptha | Download SP-120 data sheet | Use with addition or condensation cure. Available in Red. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SP-121 | 3.2 % | Naptha | Download SP-121 data sheet | Use with addition or condensation cure. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SP-124 | 10 % | Naptha | Download SP-124 data sheet | Use with addition or condensation cure. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SP-142 | 15 % | Naptha | Download SP-142 data sheet | Recommended where platinum inhibition is of concern. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SP-270 | 15 % | Naptha | Download SP-270 data sheet | Improved adhesion to gold - Recommended where platinum inhibition is of concern such as when bonding on polyphenylsulphone. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SP-271 | 20 % | Naptha | Download SP-271 data sheet | Recommended where platinum inhibition is of concern. | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
Resins
Viscosity, Durometer, Appearance,Product | Viscosity | Durometer | Appearance | Data Sheet | Comment | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CF2-4721 | 130 cP | 75 Type D | Light Amber | Download CF2-4721 data sheet | Hard Resin;Contains vinyl-specific peroxide in 1.52 RI resin. Requires heat to cure and oxygen inhibited. Not recommended for general encapsulation. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
Soft Silicone Adhesives
Tack, Viscosity, Penetration, Work Time,Product | Tack | Viscosity | Penetration | Work Time | Data Sheet | Comment | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
MED-6342 | 10 psi | 10,300 cP | 1 mm | 10 hours | Download MED-6342 data sheet | Low Viscosity, Firm Gel | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6345 | 5.8 psi | 15,300 cP | 5 mm | 30 minutes | Download MED-6345 data sheet | Medium Viscosity, Soft Gel | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6350 | 6 psi | 20,750 cP | 15 mm | 2 hours | Download MED-6350 data sheet | Medium Viscosity, Soft Gel | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6360 | 12 psi | 450 cP | 14 mm | N/A | Download MED-6360 data sheet | Very Low Viscosity, Soft Gel | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
MED-6362 | N/A | 8,500 cP | 13 mm | N/A | Download MED-6362 data sheet | Low Viscosity, Soft Gel | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
Specialty Fluids
Appearance, Viscosity, D4 Content, INCI Name,Product | Appearance | Viscosity | D4 Content | INCI Name | Data Sheet | Comment | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CSF-3504 | Clear | 1,140,000 cP | <0.05 % | Dimethicone (and) Dimethiconol | Download CSF-3504 data sheet | 50% High Purity Dimethiconol Dispersed in Low Viscosity Dimethicone Fluid | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
Specialty Polymers
Refractive Index, Viscosity, Mmol, % Pt, Appearance,Product | Refractive Index | Viscosity | Mmol | % Pt | Appearance | Data Sheet | Comment | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CAT-102 | N/A | N/A | N/A | N/A | White | Download CAT-102 data sheet | N/A | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CAT-41 | N/A | N/A | N/A | N/A | Translucent Yellow | Download CAT-41 data sheet | N/A | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CAT-50 | 1.40 | N/A | N/A | 2.5 | Light Amber | Download CAT-50 data sheet | N/A | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CAT-53 | 1.52 | N/A | N/A | 2 | Light Amber | Download CAT-53 data sheet | N/A | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CF-4721 | 1.52 | 125 cP | 4.7 Vi/g | N/A | Clear | Download CF-4721 data sheet | 75 Type D with Dicumyl Peroxide Catalyst (Catalyst Not Included) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
PLY-7511 | 1.41 | 1,000 cSt | 1.1 Vi/g | N/A | Translucent | Download PLY-7511 data sheet | N/A | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
PLY-7665 | 1.52 | 575 cSt | 0.8 Vi/g | N/A | Translucent | Download PLY-7665 data sheet | N/A | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
XL-110 | 1.40 | 4 cSt | 7 H/g | N/A | Translucent | Download XL-110 data sheet | N/A | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
XL-112 | 1.40 | 40 cSt | 6.5 H/g | N/A | Translucent | Download XL-112 data sheet | N/A | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
XL-115 | 1.40 | 50 cSt | 4.2 H/g | N/A | Translucent | Download XL-115 data sheet | N/A | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
XL-119 | N/A | N/A | 11.5 Vi/g | N/A | Clear | Download XL-119 data sheet | Compatible with a variety of silicone polymer systems for formulating silicone adhesives, elastomers and gels due to its reactivity and solubility with silicone | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
XL-128 | N/A | N/A | N/A | N/A | Translucent | Download XL-128 data sheet | N/A | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
XL-151 | 1.40 | 480 cSt | 0.4 H/g | N/A | Translucent | Download XL-151 data sheet | For crosslinking silicone addition cure polymer systems, to form adhesives, sealant, rubbers and gels due to its reactivity with vinyl functional groups | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
XL-7505 | 1.40 | 500 cP | 0.16 H/g | N/A | Translucent | Download XL-7505 data sheet | N/A | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
Thermally Conductive
Thermal Conductivity, Mix Ratio, Viscosity, Extrusion Rate, Cure System,Product | Thermal Conductivity | Mix Ratio | Viscosity | Extrusion Rate | Cure System | Data Sheet | Comment | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CF1-3800 | 1.25 W/(mK) | 15:1 | N/A | N/A | Platinum | Download CF1-3800 data sheet | 50% M Fluoro. Recommended for short term exposure to solvents and fuel. (Contains Boron Nitride) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2900 | 0.61 W/(mK) | 1 PART | N/A | 40 g/minute | Oxime | Download CV-2900 data sheet | *22) 0.6 W/mK, Low Temperature | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2942 | 1 W/(mK) | 20:1 | N/A | N/A | Platinum | Download CV-2942 data sheet | *22) 1.0 W/mK, *4) Primed Lap Shear 375 psi (2.6 MPa) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2943 | 1.22 W/(mK) | 100:0.2 | N/A | N/A | Alkoxy | Download CV-2943 data sheet | *22) 1.2 W/mK, *3) Primed Lap Shear 475 psi (3.3 MPa) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2946 | 1.49 W/(mK) | 15:1 | 3,560,000 cP | N/A | Platinum | Download CV-2946 data sheet | *22) 1.5 W/mK *,4 ) Primed Lap Shear 160 psi (1.1 MPa) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2948 | 1.95 W/(mK) | 100:0.2 | N/A | N/A | Alkoxy | Download CV-2948 data sheet | *22) 2.0 W/mK, *3) Primed Lap Shear 150 psi (1.0 MPa) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2960 | 0.83 W/(mK) | 10:1 | 130,000 cP | N/A | Platinum | Download CV-2960 data sheet | *22) 0.8 W/mK, *4) Primed Lap Shear 205 psi (1.4 MPa), Low Viscosity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2961 | 0.79 W/(mK) | 10:1 | 300,000 cP | N/A | Platinum | Download CV-2961 data sheet | *22) 0.8 W/mK Low Viscosity, *3) Primed Lap Shear 205 psi (1.4 MPa), Low Temp | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV-2963 | 0.64 W/(mK) | 20:1 | N/A | N/A | Platinum | Download CV-2963 data sheet | Primed w/CF1-135, Contains Boron Nitride | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV1-2960 | 1.11 W/(mK) | 10:1 | 900,000 cP | N/A | Platinum | Download CV1-2960 data sheet | *22) 1.1 W/mK | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV1-2964 | 0.95 W/(mK) | 1:1 | 50,000 cP | N/A | Platinum | Download CV1-2964 data sheet | *22) 1.0 W /mK, *5) Primed Lap Shear 120 psi (0.8 MPa) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV2-2946 | 0.64 W/(mK) | 20:1 | N/A | 140 g/minute | Platinum | Download CV2-2946 data sheet | *22) 0.6 W/mK, Thin Bond Line | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CV4-2946 | 0.95 W/(mK) | N/A | N/A | 122 g/minute | Platinum | Download CV4-2946 data sheet | Gel, soft and compliant | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
EPM-2490 | 1.49 W/(mK) | 15:1 | 3,672,800 cP | N/A | Platinum | Download EPM-2490 data sheet | 1.5 W/mK, 160 psi lap shear primed w/CF1-135, 2 hr. pot life. (Contains Boron Nitride) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
EPM1-2493 | 0.95 W/(mK) | 1:1 | 36,000 cP | N/A | Platinum | Download EPM1-2493 data sheet | Recommended for bondlines 5 micron or greater. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
G-9340 | 0.68 W/(mK) | N/A | N/A | N/A | Non-Curing | Download G-9340 data sheet | Versatile, thermally conductive grease where thin bondlines or a compressible grease is needed. Non-curing. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2165 | 0.5 W/(mK) | 1:1 | 4,000 cP | N/A | Platinum | Download R-2165 data sheet | 0.6 W/mK, versatile, moderate thermally conductive silicone. Will RTV. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2175 | 0.4 W/(mK) | 1:1 | 2,000 cP | N/A | Platinum | Download R-2175 data sheet | 0.4 W/mk, pourable, heat resistant, tested per UL94 and passed V0. Will RTV. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2930 | 1.46 W/(mK) | 15:1 | N/A | N/A | Platinum | Download R-2930 data sheet | Smooth Paste;1.45 W/mK, 3 hr. pot life. (Contains Boron Nitride) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2939 | 0.75 W/(mK) | 15:1 | 70,000 cP | N/A | Platinum | Download R-2939 data sheet | 0.75 W/mK, 4 hr. pot life. (Contains Boron Nitride) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2940 | 0.84 W/(mK) | 20:1 | N/A | N/A | Platinum | Download R-2940 data sheet | Smooth Paste;0.8 W/mK, 5 hr. pot life. | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R-2949 | 0.75 W/(mK) | 15:1 | 75,000 cP | N/A | Platinum | Download R-2949 data sheet | For applications requiring a broader operating temperature range. (Contains Boron Nitride) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SCV1-2599 | 1.6 W/(mK) | 15:1 | 3,750,000 cP | N/A | Platinum | Download SCV1-2599 data sheet | Primed w/CF1-135, Contains Boron Nitride | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Product Details
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SCV2-2599 | 0.64 W/(mK) | 20:1 | N/A | 140 g/minute | Platinum | Download SCV2-2599 data sheet | 0.64 W /mK (Tested per ASTM E1530) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||